Lithium nitrate, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, and rocket propellant
Purity: 99%
CAS No.: 7790-69-4
Molecular Formula: LiNO3
Formula Weight: 68.95
Physical Form: Crystalline
Density: 2.38
Appearance: White
Odor: Odorless
Melting Point: 264°
Boiling Point: 600° dec.
Solubility: Soluble in water, alcohol, NH4OH, and pyridine
Merck Reference: 14,5536
UN No.: 2722
MDL No.: MFCD00011094
Used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, and rocket propellant
Formula:
LiNO₃ MW: 68.95 g/mol Boiling Pt: 600 °C (dec.) Melting Pt: 264 °C Density: 2.38 Storage Temperature: Ambient |
MDL Number:
MFCD00011094 CAS Number: 7790-69-4 EINECS: 232-218-9 UN: 2722 ADR: 5.1,III Merck Index: 14,5536 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...