Lithium sulfate, anhydrous 99,7 Metals basis

Supplier: Thermo Scientific Chemicals
Warning

13404-30 13404-A1 13404-A7
CAAA13404-30EA 104.28 CAD
CAAA13404-30 CAAA13404-A1 CAAA13404-A7
Lithium sulfate, anhydrous 99,7 Metals basis
Lithium sulphate

MDL: MFCD00011086


Purity: 99.7% CAS No.: 10377-48-7 Molecular Formula: Li2SO4 Formula Weight: 109.94 Physical Form: -40 Mesh Powder Density: 2.22 Appearance: White Odor: Odorless Melting Point: 845° Merck Reference: 14,5541 MDL No.: MFCD00011086

Formula: Li₂O₄S
MW: 109.95 g/mol
Boiling Pt: 1377
Melting Pt: 859
Density: 2.22
Storage Temperature: Ambient
MDL Number: MFCD00011086
CAS Number: 10377-48-7
EINECS: 233-820-4
Merck Index: 14,5541

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR