Tungsten trioxide ≥99.998% (metals basis excluding Mo), Puratronic® (max. 0.01% Mo)
Supplier: Thermo Scientific Chemicals
|
Warning
|
13398-14
13398-22
CAAA13398-14EA
251.22
CAD
CAAA13398-14
CAAA13398-22
Tungsten trioxide ≥99.998% (metals basis excluding Mo), Puratronic® (max. 0.01% Mo)
Tungsten trioxide
MDL: MFCD00011466
Purity: 99.998%
CAS No.: 1314-35-8
Molecular Formula: O3W
Formula Weight: 231.85
Physical Form: Powder
Density: 7.16
Melting Point: 1473°
Merck Reference: 14,9817
MDL No.: MFCD00011466
Formula:
O₃W Melting Pt: 1473 °C Density: 7.16 Storage Temperature: Ambient |
MDL Number:
MFCD00011466 CAS Number: 1314-35-8 EINECS: 215-231-4 Merck Index: 14,9817 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...