Copper ≥99.9% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

13380-CU 13380-CV
CAAA13380-CUEA 90.53 CAD
CAAA13380-CU CAAA13380-CV
Copper ≥99.9% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Copper

MDL: MFCD00010965


Purity: 99.9% CAS No.: 7440-50-8 Physical Form: ≈45.5g/20x20cm, Temper: soft Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR