Copper ≥99.9% (metals basis), foil, Thickness: 0.254 mm (0.010 in)

Supplier: Thermo Scientific Chemicals

13379-CU 13379-CV
CAAA13379-CUEA 131.61 CAD
CAAA13379-CU CAAA13379-CV
Copper ≥99.9% (metals basis), foil, Thickness: 0.254 mm (0.010 in)
Copper

MDL: MFCD00010965


Purity: 99.9% CAS No.: 7440-50-8 Physical Form: ≈91g/20x20cm Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR