Copper ≥99.9% (metals basis), foil, Thickness: 1.27 mm (0.05 in)

Supplier: Thermo Scientific Chemicals

13377-CU 13377-CV
CAAA13377-CUEA 284.4 CAD
CAAA13377-CU CAAA13377-CV
Copper ≥99.9% (metals basis), foil, Thickness: 1.27 mm (0.05 in)
Copper

MDL: MFCD00010965


Purity: 99.9% CAS No.: 7440-50-8 Physical Form: ≈455g/20x20cm Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR