Arsenic ≥99.9999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
13375-09
13375-14
13375-22
13375-A1
CAAA13375-03EA
144.74
CAD
CAAA13375-03
CAAA13375-09
CAAA13375-14
CAAA13375-22
CAAA13375-A1
Arsenic ≥99.9999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Arsenic
MDL: MFCD00085309
Purity: 99.9999%
CAS No.: 7440-38-2
Physical Form: Ampouled under argon
Odor: Odorless
UN No.: 1558
MDL No.: MFCD00085309
Formula:
As Melting Pt: 817 °C Density: 5.727 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085309 CAS Number: 7440-38-2 EINECS: 213-148-6 UN: 1558 ADR: 6.1,II Merck Index: 12,00832 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...