Hafnium ≥99.7% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

13305-BU 13305-BY
CAAA13305-BUEA 152 CAD
CAAA13305-BU CAAA13305-BY CAAA13305-CC
Hafnium ≥99.7% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 2.0 mm (0.08 in)
Hafnium

MDL: MFCD00011032


Purity: 99.7% CAS No.: 7440-58-6 Physical Form: ≈41.1g/m Odor: Odorless MDL No.: MFCD00011032

Formula: Hf
MW: 178.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR