Molybdenum ≥99.7% (metals basis), pellets

Supplier: Thermo Scientific Chemicals

13242-22 13242-36 13242-A3
CAAA13242-22EA 88.16 CAD
CAAA13242-22 CAAA13242-36 CAAA13242-A3
Molybdenum ≥99.7% (metals basis), pellets
Molybdenum

MDL: MFCD00003465


Purity: 99.7% CAS No.: 7439-98-7 Physical Form: ≈12.5x18x25mm, Hot Pressed, density ≈8.0g/cm3 MDL No.: MFCD00003465

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR