Zinc selenide ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc (II) selenide

13241-18 13241-30
CAAA13241-18EA 287.56 CAD
CAAA13241-18 CAAA13241-30
Zinc selenide ≥99.995% (metals basis)
Zinc selenide

MDL: MFCD00011296


Purity: 99.995% CAS No.: 1315-09-9 Molecular Formula: ZnSe Formula Weight: 144.33 Physical Form: 3-12mm Lump Density: 5.42 Melting Point: 1100° Merck Reference: 14,10156 UN No.: 3283 MDL No.: MFCD00011296

Formula: SeZn
MW: 144.35 g/mol
Melting Pt: >1100 °C
Density: 5.42
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1 III
Merck Index: 14,10156

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR