Cesium hydroxide hydrate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cesium hydroxide, hydrate

13233-06 13233-14 13233-22
CAAA13233-06EA 55.14 CAD
CAAA13233-06 CAAA13233-14 CAAA13233-22
Cesium hydroxide hydrate ≥99.9% (metals basis)
Cesium hydroxide hydrate

MDL: MFCD00010964 Fieser: 21,96


Purity: 99.9% CAS No.: 12260-45-6 Molecular Formula: CsOH.xH2O (15-20% H2O) Formula Weight: 149.91(anhy) Physical Form: Crystalline Sensitivity: Air Sensitive and Hygroscopic Appearance: White Odor: Odorless Merck Reference: 14,2013 UN No.: 2682 MDL No.: MFCD00010964

Formula: HCsO·H₂O
MW: 167.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010964
CAS Number: 12260-45-6
EINECS: 244-344-1
UN: 2682
ADR: 8,II
Merck Index: 14,2013

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR