Rhenium trichloride

Supplier: Thermo Scientific Chemicals
Warning

13230-01 13230-03
CAAA13230-01EA 268.6 CAD
CAAA13230-01 CAAA13230-03
Rhenium trichloride
Rhenium trichloride

MDL: MFCD00011196


CAS No.: 13569-63-6 Molecular Formula: ReCl3 Formula Weight: 292.56 Physical Form: Powder Sensitivity: Moisture Sensitive Odor: Odorless MDL No.: MFCD00011196

Formula: Cl₃Re
Storage Temperature: Ambient
MDL Number: MFCD00011196
CAS Number: 13569-63-6
EINECS: 236-987-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR