Terbium(III) bromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
13173-09
13173-18
CAAA13173-09EA
154.68
CAD
CAAA13173-09
CAAA13173-18
Terbium(III) bromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Terbium (III) bromide hydrate
MDL: MFCD00049571
Purity: 99.99%
Molecular Formula: TbBr3.xH2O
Formula Weight: 398.65(anhy)
Physical Form: Crystalline
Melting Point: 170° dec.
MDL No.: MFCD00049571
Formula:
Br₃Tb·H₂O Melting Pt: 170 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00049571 CAS Number: 15162-98-8 EINECS: 238-442-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...