Yttrium(III) bromide, hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

13158-14 13158-22
CAAA13158-14EA 161.16 CAD
CAAA13158-14 CAAA13158-22 CAAA13158-30
Yttrium(III) bromide, hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Yttrium (III) bromide

MDL: MFCD00049710


Purity: 99.99% Molecular Formula: YBr3.xH2O Formula Weight: 328.63(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic


Caution: Hygroscopic. Store in cool, dry, conditions in well sealed container. Incompatible with water, moisture.

Formula: YBr₃·xH₂O
MW: 328.62
Storage Temperature: Ambient
MDL Number: MFCD00049710
CAS Number: 13469-98-2
EINECS: 236-728-2

Order Now

Specification Test Results

Total Rare Earth Oxide impurities 0.01% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR