Nitronium hexafluorophosphate

Supplier: Thermo Scientific Chemicals
Danger

13151-06 13151-14
CAAA13151-06EA 219.62 CAD
CAAA13151-06 CAAA13151-14
Nitronium hexafluorophosphate
Nitronium hexafluorophosphate

MDL: MFCD00031520


CAS No.: 19200-21-6 Molecular Formula: NO2PF6 Formula Weight: 190.97 Physical Form: Crystalline Sensitivity: Moisture Sensitive MDL No.: MFCD00031520

Formula: F₆NO₂P
Storage Temperature: Ambient
MDL Number: MFCD00031520
CAS Number: 19200-21-6
EINECS: 242-872-7
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR