Nitrosium hexafluorophosphate ≥96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nitrosonium hexafluorophosphate, Nitrosyl hexafluorophosphate

13149-06 13149-14
CAAA13149-06EA 218.04 CAD
CAAA13149-06 CAAA13149-14
Nitrosium hexafluorophosphate ≥96%
Nitrosium hexafluorophosphate

MDL: MFCD00040324


Purity: 96% CAS No.: 16921-91-8 Molecular Formula: F6NOP Formula Weight: 174.96 Physical Form: Crystalline Sensitivity: Moisture Sensitive UN No.: 3260 MDL No.: MFCD00040324

Formula: F₆NOP
MW: 174.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040324
CAS Number: 16921-91-8
EINECS: 240-980-9
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR