Potassium peroxodisulfate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
13145-30
13145-A1
CAAA13145-30EA
92.27
CAD
CAAA13145-30
CAAA13145-A1
Potassium peroxodisulfate ≥99.0% ACS
Potassium peroxodisulphate
A polymerization accelerant for PAGE gels
Purity: 99.0% min
CAS No.: 7727-21-1
Molecular Formula: K2S2O8
Formula Weight: 270.31
Physical Form: Crystalline
Density: 2.477
Melting Point: 100° dec.
Merck Reference: 14,7656
UN No.: 1492
MDL No.: MFCD00011386
A polymerization accelerant for PAGE gels
Formula:
K₂S₂O₈ Melting Pt: 100 °C (dec.) Density: 2.477 Storage Temperature: Ambient |
MDL Number:
MFCD00011386 CAS Number: 7727-21-1 EINECS: 231-781-8 UN: 1492 ADR: 5.1,III Merck Index: 14,7656 |
Specification Test Results
Insoluble matter | 0.005% |
Chlorine compounds (as Cl) | 0.001% |
Heavy Metals (as Pb) | 0.001% |
Fe | 5 ppm |
Mn | 2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...