Manganese(IV) telluride ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

13134-09 13134-18
CAAA13134-09EA 157.84 CAD
CAAA13134-09 CAAA13134-18
Manganese(IV) telluride ≥99.9% (metals basis)
Manganese (IV) telluride

MDL: MFCD00064703


Purity: 99.9% CAS No.: 12032-89-2 Molecular Formula: MnTe2 Formula Weight: 310.14 Physical Form: 6mm & Down Pieces Odor: Odorless MDL No.: MFCD00064703

Formula: MnTe₂
Storage Temperature: Ambient
MDL Number: MFCD00064703
CAS Number: 12032-89-2
EINECS: 234-783-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR