Diantimony triselenide ≥99.999% (metals basis), pieces
Supplier: Thermo Scientific Chemicals
|
Danger
|
13130-06
13130-14
CAAA13130-06EA
248.06
CAD
CAAA13130-06
CAAA13130-14
Diantimony triselenide ≥99.999% (metals basis), pieces
Diantimony triselenide
MDL: MFCD00016319
Very slightly soluble in water.
Purity: 99.999%
CAS No.: 1315-05-5
Molecular Formula: Sb2Se3
Formula Weight: 480.38
Physical Form: 3-12mm Pieces
Sensitivity: Moisture Sensitive
Solubility: Very slightly soluble in water
Merck Reference: 14,712
UN No.: 3077
MDL No.: MFCD00016319
Formula:
Sb₂Se₃ MW: 480.4 g/mol Melting Pt: 611 °C Density: 5.81 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016319 CAS Number: 1315-05-5 EINECS: 215-256-0 UN: 3077 ADR: 9,III Merck Index: 14,712 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...