Dibismuth triselenide ≥99.999% (metals basis) for vacuum deposition

Supplier: Thermo Scientific Chemicals
Danger

13126-09 13126-18
CAAA13126-09EA 145.2 CAD
CAAA13126-09 CAAA13126-18
Dibismuth triselenide ≥99.999% (metals basis) for vacuum deposition
Dibismuth triselenide

In semiconductor research, multilayer thin-film, magnetoresistant film


Purity: 99.999% CAS No.: 12068-69-8 Molecular Formula: Bi2Se3 Formula Weight: 654.84 Physical Form: 1-6 mm Pieces Density: 6.82 Appearance: Grey Melting Point: 710° Solubility: Insoluble in water. Decomposes when heated in air. Decomposes in nitric acid and aqua regia Merck Reference: 14,1278 UN No.: 3283 MDL No.: MFCD00014200


In semiconductor research, multilayer thin-film, magnetoresistant film

Formula: Bi₂Se₃
MW: 654.84 g/mol
Melting Pt: 710 °C
Density: 6.82
Storage Temperature: Ambient
MDL Number: MFCD00014200
CAS Number: 12068-69-8
EINECS: 235-104-7
UN: 3283
ADR: 6.1,III
Merck Index: 14,1278

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR