Cadmium(II) iodide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

13125-14 13125-22
CAAA13125-14EA 141.09 CAD
CAAA13125-14 CAAA13125-22
Cadmium(II) iodide ≥99.5% (metals basis)
Cadmium (II) iodide

In photography, process engraving and lithography, analytical chemistry, electrodeposition of Cadmium, manufacturing of phosphors, lubricants


Purity: 99.5% CAS No.: 7790-80-9 Molecular Formula: CdI2 Formula Weight: 366.21 Physical Form: -40 Mesh Density: 5.67 Appearance: Green-yellow Odor: Odorless Melting Point: 387-389° Boiling Point: 787° Solubility: Soluble in water, alcohol, ether, acetone, ammonia, and acids Merck Reference: 14,1621 UN No.: 2570 MDL No.: MFCD00010920


In photography, process engraving and lithography, analytical chemistry, electrodeposition of Cadmium, manufacturing of phosphors, lubricants

Formula: CdI₂
Boiling Pt: 787 °C
Melting Pt: 387…389 °C
Density: 5.67
Storage Temperature: Ambient
MDL Number: MFCD00010920
CAS Number: 7790-80-9
EINECS: 232-223-6
UN: 2570
ADR: 6.1,III
Merck Index: 14,1621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR