Cobalt sulfide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

13114-14 13114-22
CAAA13114-14EA 167.48 CAD
CAAA13114-14 CAAA13114-22
Cobalt sulfide ≥99.5% (metals basis)
Cobalt sulphide

MDL: MFCD00016032


Purity: 99.5% CAS No.: 1317-42-6 Molecular Formula: CoS (generally Co6S5 and/or Co4S3) Formula Weight: 91.00 Physical Form: -100 Mesh Powder Odor: Odorless UN No.: 3077 MDL No.: MFCD00016032

Formula: CoS
Melting Pt: >1116 °C
Density: 5.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016032
CAS Number: 1317-42-6
EINECS: 215-273-3
UN: 3077
ADR: 9,III
Merck Index: 12,02511

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR