Molybdenum diselenide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

13112-06 13112-14
CAAA13112-06EA 153.26 CAD
CAAA13112-06 CAAA13112-14
Molybdenum diselenide ≥99.9% (metals basis)
Molybdenum diselenide

MDL: MFCD00014216


Purity: 99.9% CAS No.: 12058-18-3 Molecular Formula: MoSe2 Formula Weight: 253.87 Physical Form: -325 Mesh Powder Odor: Odorless UN No.: 3077 MDL No.: MFCD00014216

Formula: MoSe₂
Melting Pt: >1200 °C
Density: 6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014216
CAS Number: 12058-18-3
EINECS: 235-027-9
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR