Nickel(II) telluride ≥99.9% (metals basis excluding Co) (max. 0.1-1% Co)

Supplier: Thermo Scientific Chemicals
Danger

13105-09 13105-18
CAAA13105-09EA 127.51 CAD
CAAA13105-09 CAAA13105-18
Nickel(II) telluride ≥99.9% (metals basis excluding Co) (max. 0.1-1% Co)
Nickel (II) telluride

MDL: MFCD00016258


Purity: 99.9% CAS No.: 12142-88-0 Molecular Formula: NiTe Formula Weight: 186.31 Physical Form: 10mm & Down Lump Odor: Odorless UN No.: 3077 MDL No.: MFCD00016258

Formula: NiTe
Storage Temperature: Ambient
MDL Number: MFCD00016258
CAS Number: 12142-88-0
EINECS: 235-260-6
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR