Selenium tetrachloride ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

13096-14 13096-22
CAAA13096-14EA 194.34 CAD
CAAA13096-14 CAAA13096-22
Selenium tetrachloride ≥99.5% (metals basis)
Selenium tetrachloride

MDL: MFCD00011225 Decomposes in water and moist air. Insoluble in liquid bromine. Sublimes when heated


Purity: 99.5% CAS No.: 10026-03-6 Molecular Formula: SeCl4 Formula Weight: 220.77 Physical Form: Lump Density: 2.600 Refractive Index: 1.807 Sensitivity: Moisture Sensitive Odor: Pungent Melting Point: subl. Solubility: Decomposes in water and moist air. Insoluble in liquid bromine. Sublimes when heated Merck Reference: 14,8438 UN No.: 2923 MDL No.: MFCD00011225

Formula: Cl₄Se
Melting Pt: (subl.)
Density: 2.600
Storage Temperature: Ambient
MDL Number: MFCD00011225
CAS Number: 10026-03-6
EINECS: 233-053-5
UN: 2923
ADR: 8,II
Merck Index: 14,8438

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR