Tungsten diselenide ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

13084-09 13084-18
CAAA13084-09EA 153.42 CAD
CAAA13084-09 CAAA13084-18
Tungsten diselenide ≥99.8% (metals basis)
Tungsten diselenide

MDL: MFCD00049703


Purity: 99.8% CAS No.: 12067-46-8 Molecular Formula: WSe2 Formula Weight: 341.77 Physical Form: 10-20 Micron powder Density: 9.32 Appearance: Black Odor: Odorless UN No.: 3077 MDL No.: MFCD00049703

Formula: Se₂W
Density: 9,32
Storage Temperature: Ambient
MDL Number: MFCD00049703
CAS Number: 12067-46-8
EINECS: 235-078-7
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR