Titanium(III) fluoride

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Titanium trifluoride

13079-09 13079-18
CAAA13079-09EA 107.44 CAD
CAAA13079-09 CAAA13079-18
Titanium(III) fluoride
Titanium (III) fluoride

MDL: MFCD00042544


CAS No.: 13470-08-1 Molecular Formula: TiF3 Formula Weight: 104.90 Physical Form: Powder Density: 3.400 Sensitivity: Air and Moisture Sensitive UN No.: 3260 MDL No.: MFCD00042544

Formula: F₃Ti
MW: 104.88
Density: 3.400
Storage Temperature: Ambient
MDL Number: MFCD00042544
CAS Number: 13470-08-1
EINECS: 236-732-4
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR