Copper(II) fluoride dihydrate

Supplier: Thermo Scientific Chemicals
Danger

13076-18 13076-30
CAAA13076-18EA 84.85 CAD
CAAA13076-18 CAAA13076-30
Copper(II) fluoride dihydrate
Copper (II) fluoride dihydrate

Flux in casting gray iron


CAS No.: 13454-88-1 Molecular Formula: CuF2.2H2O Formula Weight: 137.57 (101.54anhy) Physical Form: Powder Density: 2.934 Odor: Odorless Merck Reference: 14,2638 UN No.: 3288 MDL No.: MFCD00010977


Flux in casting gray iron

Formula: CuF₂·2H₂O
Melting Pt: >130 °C (dec.)
Density: 2.934
Storage Temperature: Ambient
MDL Number: MFCD00010977
CAS Number: 13454-88-1
EINECS: 232-147-3
UN: 3288
ADR: 6.1,III
Merck Index: 14,2638

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR