Bismuth(III) fluoride, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
13075-14
13075-22
13075-36
CAAA13075-14EA
83.9
CAD
CAAA13075-14
CAAA13075-22
CAAA13075-36
Bismuth(III) fluoride, anhydrous 99%
Bismuth (III) fluoride
Manufacturing of bismuth pentafluoride
Purity: 99%
CAS No.: 7787-61-3
Molecular Formula: BiF3
Formula Weight: 265.98
Physical Form: Powder
Refractive Index: 1.74
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 727°
Solubility: Practically insoluble in water. Soluble in HF
Merck Reference: 14,1265
UN No.: 3260
MDL No.: MFCD00010893
Manufacturing of bismuth pentafluoride
Formula:
BiF₃ Boiling Pt: 230 °C (1013 hPa) Melting Pt: 727 °C Density: 3.7 g/cm³ (160 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010893 CAS Number: 7787-61-3 EINECS: 232-124-8 UN: 3260 ADR: 8,III Merck Index: 14,1265 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...