Cobalt(II) fluoride, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt difluoride, Cobaltous fluoride

13074-14 13074-22
CAAA13074-14EA 135.25 CAD
CAAA13074-14 CAAA13074-22
Cobalt(II) fluoride, anhydrous 98%
Cobalt (II) fluoride

MDL: MFCD00010941


Purity: 98% CAS No.: 10026-17-2 Molecular Formula: CoF2 Formula Weight: 96.93 Physical Form: Powder Density: 4.46 Melting Point: 1200° Boiling Point: 1400° Merck Reference: 14,2440 UN No.: 2923 MDL No.: MFCD00010941

Formula: CoF₂
MW: 96.93
Boiling Pt: 1400
Melting Pt: 1200
Density: 4.46
Storage Temperature: Ambient
MDL Number: MFCD00010941
CAS Number: 10026-17-2
EINECS: 233-061-9
UN: 2923
ADR: 8 III
Merck Index: 14,2440

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR