Manganese(III) fluoride ≥98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
13069-09
13069-18
CAAA13069-09EA
82.48
CAD
CAAA13069-09
CAAA13069-18
Manganese(III) fluoride ≥98% (metals basis)
Manganese (III) fluoride
MDL: MFCD00016223
Purity: 98%
CAS No.: 7783-53-1
Molecular Formula: MnF3
Formula Weight: 111.93
Physical Form: Crystalline
Density: 3.54
Melting Point: dec.
Merck Reference: 14,5741
UN No.: 3288
MDL No.: MFCD00016223
Formula:
F₃Mn MW: 111.93 g/mol Melting Pt: Dec. Density: 3.54 Storage Temperature: Ambient |
MDL Number:
MFCD00016223 CAS Number: 7783-53-1 EINECS: 232-006-6 UN: 3087 ADR: 5.1,II Merck Index: 14,5741 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...