Iron(III) fluoride trihydrate

Supplier: Thermo Scientific Chemicals
Danger

13068-18 13068-30
CAAA13068-18EA 126.72 CAD
CAAA13068-18 CAAA13068-30
Iron(III) fluoride trihydrate
Iron (III) fluoride trihydrate

MDL: MFCD00149718


CAS No.: 15469-38-2 Molecular Formula: FeF3.3H2O Formula Weight: 166.89 (112.84anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Off-white to pale brown Merck Reference: 14,4022 UN No.: 3260 MDL No.: MFCD00149718

Formula: F₃Fe·3H₂O
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149718
CAS Number: 15469-38-2
EINECS: 232-002-4
UN: 3260
ADR: 8,II
Merck Index: 14,4022

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR