Nickel(II) fluoride, anhydrous 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
13067-09
13067-18
CAAA13067-09EA
122.92
CAD
CAAA13067-09
CAAA13067-18
Nickel(II) fluoride, anhydrous 97%
Nickel (II) fluoride
MDL: MFCD00016263
Slightly soluble in water
Purity: 97%
CAS No.: 10028-18-9
Molecular Formula: NiF2
Formula Weight: 96.71
Physical Form: Crystalline
Density: 4.72
Appearance: Pale yellow
Odor: Odorless
Solubility: Slightly soluble in water
Merck Reference: 14,6508
UN No.: 3288
MDL No.: MFCD00016263
Formula:
F₂Ni Melting Pt: 1360…1380 °C Density: 4.72 Storage Temperature: Ambient |
MDL Number:
MFCD00016263 CAS Number: 10028-18-9 EINECS: 233-071-3 UN: 3288 ADR: 6.1,III Merck Index: 14,6508 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...