Sodium hexafluoroantimonate ≥99.5%

Supplier: Thermo Scientific Chemicals
Warning

13066-14 13066-22
CAAA13066-14EA 94.33 CAD
CAAA13066-14 CAAA13066-22
Sodium hexafluoroantimonate ≥99.5%
Sodium hexafluoroantimonate

MDL: MFCD00003482


Purity: 99.5% CAS No.: 16925-25-0 Molecular Formula: NaSbF6 Formula Weight: 258.73 Physical Form: Crystalline Density: 3.38 UN No.: 1549 MDL No.: MFCD00003482

Formula: F₆NaSb
Density: 3,38
Storage Temperature: Ambient
MDL Number: MFCD00003482
CAS Number: 16925-25-0
EINECS: 240-989-8
UN: 1549
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR