Sodium hexafluorophosphate 99+%

Supplier: Thermo Scientific Chemicals
Danger

13064-14 13064-22
CAAA13064-14EA 211.72 CAD
CAAA13064-14 CAAA13064-22
Sodium hexafluorophosphate 99+%
Sodium hexafluorophosphate

MDL: MFCD00011122


Purity: 99+% CAS No.: 21324-39-0 Molecular Formula: NaPF6 Formula Weight: 167.95 Physical Form: Crystalline Density: 2.37 Melting Point: 200° UN No.: 3260 MDL No.: MFCD00011122

Formula: F₆NaP
Melting Pt: >200 °C
Density: 2.37
Storage Temperature: Ambient
MDL Number: MFCD00011122
CAS Number: 21324-39-0
EINECS: 244-333-1
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR