Tin(IV) fluoride ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
13063-04
13063-09
CAAA13063-04EA
126.08
CAD
CAAA13063-04
CAAA13063-09
Tin(IV) fluoride ≥99% (metals basis)
Tin (IV) fluoride
Friedel-Crafts catalyst
Purity: 99%
CAS No.: 7783-62-2
Molecular Formula: SnF4
Formula Weight: 194.68
Physical Form: -6 Mesh Crystalline
Sensitivity: Moisture Sensitive
Merck Reference: 14,8776
UN No.: 2923
MDL No.: MFCD00042541
Friedel-Crafts catalyst
Formula:
SnF₄ MW: 194.7 Boiling Pt: 705 Melting Pt: 705 °C (subl.) Density: 4.78 g/cm³ (19 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042541 CAS Number: 7783-62-2 EINECS: 232-016-0 UN: 2923 ADR: 8 II Merck Index: 14,8776 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...