Chromium hexacarbonyl 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
13061-03
13061-09
13061-18
CAAA13061-03EA
78.53
CAD
CAAA13061-03
CAAA13061-09
CAAA13061-18
Chromium hexacarbonyl 99%
Chromium hexacarbonyl
MDL: MFCD00010945
Fieser: 5,142 6,125 7,71 8,110 9,117 10,100 11,131 13,51 18,105
Purity: 99%
CAS No.: 13007-92-6
Molecular Formula: C6CrO6
Formula Weight: 220.06
Physical Form: Crystalline
Density: 1.77
Appearance: White
Melting Point: ca 152° dec.
UN No.: 3466
MDL No.: MFCD00010945
Formula:
C₆CrO₆ Boiling Pt: 210 °C (1013 hPa) Melting Pt: ∼ 152 °C (dec.) Density: 1.77 Storage Temperature: Ambient |
MDL Number:
MFCD00010945 CAS Number: 13007-92-6 EINECS: 235-852-4 UN: 3466 ADR: 6.1,III Merck Index: 13,02253 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...