Arsenic ≥99.99999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
13044-03
13044-09
13044-14
13044-22
13044-A1
CAAA13044-03EA
40.92
CAD
CAAA13044-03
CAAA13044-09
CAAA13044-14
CAAA13044-22
CAAA13044-A1
Arsenic ≥99.99999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Arsenic
Purity: 99.99999+%
CAS No.: 7440-38-2
Molecular Formula: As
Formula Weight: 74.92
Physical Form: Packaged under argon
Sensitivity: Air Sensitive
Appearance: Grey
Odor: Odorless
UN No.: 1558
MDL No.: MFCD00085309
Formula:
As Melting Pt: 817 °C Density: 5.727 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085309 CAS Number: 7440-38-2 EINECS: 213-148-6 UN: 1558 ADR: 6.1,II Merck Index: 12,00832 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...