Barium oxalate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
13009-06
13009-14
CAAA13009-06EA
58.93
CAD
CAAA13009-06
CAAA13009-14
Barium oxalate ≥99.999% (metals basis), Puratronic®
Barium oxalate
MDL: MFCD00054429
Purity: 99.999%
CAS No.: 516-02-9
Molecular Formula: C2BaO4
Formula Weight: 225.36
Physical Form: Powder
Density: 2.658
Appearance: White
Odor: Odorless
Melting Point: 400° dec.
Merck Reference: 14,985
UN No.: 1564
MDL No.: MFCD00054429
Formula:
C₂BaO₄ Melting Pt: 400 °C (dec.) Density: 2.658 Storage Temperature: Ambient |
MDL Number:
MFCD00054429 CAS Number: 516-02-9 EINECS: 208-216-9 UN: 1564 ADR: 6.1,II Merck Index: 14,985 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...