Calcium oxalate monohydrate ≥99.9985% (metals basis) (excluding 25 ppm max alkali earths), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Oxalic acid calcium salt monohydrate, Calcium ethanedioate monohydrate

13007-06 13007-14
CAAA13007-06EA 93.22 CAD
CAAA13007-06 CAAA13007-14
Calcium oxalate monohydrate ≥99.9985% (metals basis) (excluding 25 ppm max alkali earths), Puratronic®
Calcium oxalate monohydrate

MDL: MFCD00150039 Notes: - H2O at 200°C


Purity: 99.9985% CAS No.: 5794-28-5 Molecular Formula: CaC2O4.H2O Formula Weight: 146.11 (128.10anhy) Physical Form: Powder Density: 2.12 Melting Point: 200° Merck Reference: 14,1685 UN No.: 3288 MDL No.: MFCD00150039

Formula: C₂CaO₄·1H₂O
MW: 146.11 g/mol
Melting Pt: 200
Density: 2.12
Storage Temperature: Ambient
MDL Number: MFCD00150039
CAS Number: 5794-28-5
EINECS: 209-260-1
UN: 3288
ADR: 6.1 III
Merck Index: 14,1685

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR