Mercury (II) sulfide, black ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

12992-06 12992-14
CAAA12992-06EA 325.48 CAD
CAAA12992-06 CAAA12992-14
Mercury (II) sulfide, black ≥99.999% (metals basis), Puratronic®
Mercury (II) sulphide red

MDL: MFCD00011046


Purity: 99.999% CAS No.: 1344-48-5 Molecular Formula: HgS Formula Weight: 232.65 Physical Form: Powder Density: 7.70 Appearance: Black Odor: Odorless Melting Point: 583.5° subl. Merck Reference: 14,5888 MDL No.: MFCD00011046

Formula: HgS
Melting Pt: 583.5 °C (subl.)
Density: 7.70
Storage Temperature: Ambient
MDL Number: MFCD00011046
CAS Number: 1344-48-5
EINECS: 215-696-3
UN: 2025
ADR: 6.1,II
Merck Index: 14,5888

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR