Zinc fluoride ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
12986-06
12986-14
12986-22
CAAA12986-06EA
132.72
CAD
CAAA12986-06
CAAA12986-14
CAAA12986-22
Zinc fluoride ≥99.995% (metals basis), Puratronic®
Zinc fluoride
MDL: MFCD00011298
Purity: 99.995%
CAS No.: 7783-49-5
Molecular Formula: F2Zn
Formula Weight: 103.37
Physical Form: Powder
Density: 4.95
Appearance: White
Melting Point: 872°
Boiling Point: 1500°
Merck Reference: 14,10136
MDL No.: MFCD00011298
Formula:
F₂Zn Boiling Pt: 1500 °C Melting Pt: 872 °C Density: 4.95 Storage Temperature: Ambient |
MDL Number:
MFCD00011298 CAS Number: 7783-49-5 EINECS: 232-001-9 UN: 3288 ADR: 6.1,III Merck Index: 14,10136 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...