Lithium sulfate hydrate ≥99.996% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
12983-06
12983-14
CAAA12983-06EA
65.73
CAD
CAAA12983-06
CAAA12983-14
Lithium sulfate hydrate ≥99.996% (metals basis), Puratronic®
Lithium sulfate hydrate
MDL: MFCD00011086
Purity: 99.996%
CAS No.: 10377-48-7
Molecular Formula: Li2O4S.xH2O
Formula Weight: 109.94(anhy)
Physical Form: Crystalline
Density: 2.06
Appearance: White
Melting Point: 130° -H2O
Merck Reference: 14,5541
MDL No.: MFCD00011086
Formula:
Li₂O₄S·xH₂O MW: 109.9 g/mol (anhy) Melting Pt: 130 °C (-H₂O) Density: 2.06 Storage Temperature: Ambient |
MDL Number:
MFCD00011086 CAS Number: 10377-48-7 EINECS: 233-820-4 Merck Index: 14,5541 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...