Lithium acetate hydrate ≥99.998% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

12982-06 12982-14
CAAA12982-06EA 73.15 CAD
CAAA12982-06 CAAA12982-14
Lithium acetate hydrate ≥99.998% (metals basis), Puratronic®
Lithium acetate hydrate

MDL: MFCD00013057


Purity: 99.998% CAS No.: 546-89-4 Molecular Formula: LiOOCCH3.xH2O Formula Weight: 65.98(anhy) Physical Form: Crystalline Melting Point: 57.8° MDL No.: MFCD00013057

Formula: H₃CCOOLi·xH₂O
MW: 65.98 g/mol (anhy)
Melting Pt: 291 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013057
CAS Number: 546-89-4
EINECS: 208-914-3
Merck Index: 14,5521

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR