Ammonium bromide ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

12981-06 12981-14 12981-22
CAAA12981-06EA 75.05 CAD
CAAA12981-06 CAAA12981-14 CAAA12981-22
Ammonium bromide ≥99.999% (metals basis), Puratronic®
Ammonium bromide

MDL: MFCD00011419 Fieser: 6,473


Purity: 99.999% CAS No.: 12124-97-9 Molecular Formula: NH4Br Formula Weight: 97.97 Physical Form: Crystalline Density: 2.429 Appearance: White Melting Point: 452° subl. Merck Reference: 14,505 MDL No.: MFCD00011419

Formula: H₄BrN
Melting Pt: 452 °C (subl.)
Density: 2.429
Storage Temperature: Ambient
MDL Number: MFCD00011419
CAS Number: 12124-97-9
EINECS: 235-183-8
Merck Index: 14,505

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR