Calcium acetate xhydrate ≥99.9965% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
12974-06
12974-14
CAAA12974-06EA
122.77
CAD
CAAA12974-06
CAAA12974-14
Calcium acetate xhydrate ≥99.9965% (metals basis), Puratronic®
Calcium acetate xhydrate
MDL: MFCD00012448
Beilstein Registry No.: 3692527
Purity: 99.9965%
CAS No.: 114460-21-8
Molecular Formula: Ca(OOCCH3)2.xH2O
Formula Weight: 158.17(anhy)
Physical Form: Powder
Density: 1.5
Melting Point: dec.
Beilstein Reference: 3692527
Merck Reference: 14,1646
MDL No.: MFCD00012448
Formula:
(H₃CCOO)₂Ca·nH₂O Melting Pt: Dec. Density: 1.5 Storage Temperature: Ambient |
MDL Number:
MFCD00150019 CAS Number: 114460-21-8 EINECS: 200-540-9 Merck Index: 14,1646 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...