Iron(II) sulfate heptahydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Iron vitriol, Ferrous sulphate heptahydrate, Ferrous sulfate heptahydrate

12967-06 12967-14
CAAA12967-06EA 126.08 CAD
CAAA12967-06 CAAA12967-14
Iron(II) sulfate heptahydrate ≥99.999% (metals basis), Puratronic®
Iron (II) sulphate heptahydrate

MDL: MFCD00149719


Purity: 99.999% CAS No.: 7782-63-0 Molecular Formula: FeO4S.7H2O Formula Weight: 278.02 (151.91anhy) Physical Form: Crystalline Density: 1.897 Appearance: Pale green Melting Point: 64° -6H2O Merck Reference: 14,4057 MDL No.: MFCD00149719

Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64
Density: 1.897
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Merck Index: 14,4057

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR