Nickel ≥99.999% (metals basis), powder, Puratronic® (max. 100ppm C)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12966-14
12966-22
CAAA12966-14EA
259.12
CAD
CAAA12966-14
CAAA12966-22
Nickel ≥99.999% (metals basis), powder, Puratronic® (max. 100ppm C)
Nickel
MDL: MFCD00011137
Purity: 99.999%
CAS No.: 7440-02-0
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011137
Formula:
Ni Boiling Pt: 2371 °C (1013 hPa) Melting Pt: 1455 °C Density: 8.9 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...