Nickel ≥99.97% (metals basis), lumps low oxygen, vacuum remelted

Supplier: Thermo Scientific Chemicals
Danger

12957-14 12957-22
CAAA12957-14EA 429.76 CAD
CAAA12957-14 CAAA12957-22
Nickel ≥99.97% (metals basis), lumps low oxygen, vacuum remelted
Nickel

MDL: MFCD00011137


Purity: 99.97% CAS No.: 7440-02-0 Physical Form: Oxygen and Nitrogen analysis included Odor: Odorless MDL No.: MFCD00011137

Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR