Gallium antimonide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
12933-03
12933-06
CAAA12933-03EA
82.79
CAD
CAAA12933-03
CAAA12933-06
Gallium antimonide ≥99.99% (metals basis)
Gallium antimonide
MDL: MFCD00016101
Purity: 99.99%
CAS No.: 12064-03-8
Molecular Formula: GaSb
Formula Weight: 191.47
Physical Form: 6mm & down Pieces
Odor: Odorless
UN No.: 1549
MDL No.: MFCD00016101
Formula:
GaSb Melting Pt: 710 °C Density: 5.5…5.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016101 CAS Number: 12064-03-8 EINECS: 235-058-8 UN: 1549 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...